Learn More
anti-Ly-6C, Biotin, Clone: HK1.4, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128048
Catalog No. NBP128048
Add to cart
Description
Description
Ly-6C Monoclonal antibody specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry-Frozen, In vitro assay.
Specifications
Specifications
Ly-6C
HK1.4
Biotin
PBS with 0.05% Sodium Azide
Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C
Rat
IgG2c
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Murine Ly-6C, Mr 18 kDa
Flow Cytometry, Immunohistochemistry (Frozen), In vitro assay
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay
Purified
LY6C1
L3 cloned CTL cells
Protein A or G purified
RUO
Primary
17067
Mouse