Learn More
anti-Ly-6C, DyLight 488, Clone: HK1.4, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128046G
Catalog No. NBP128046G
Add to cart
Description
Description
Ly-6C Monoclonal antibody specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Ly-6C
HK1.4
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
LY6C1
L3 cloned CTL cells
Protein A or G purified
RUO
Primary
17067
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C
Rat
IgG2c
0.25 ml
Store at 4C in the dark.
Monoclonal
Murine Ly-6C, Mr 18 kDa