Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ly-6C Antibody (HK1.4), PE, Novus Biologicals™
Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP128046PE
Description
Ly-6C Monoclonal specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry.Specifications
Ly-6C | |
Monoclonal | |
PE | |
Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C | |
Rat | |
Protein A or G purified | |
RUO | |
17067 | |
Mouse | |
Purified |
Flow Cytometry | |
HK1.4 | |
Flow Cytometry | |
LY6C1 | |
L3 cloned CTL cells | |
0.25 mL | |
Primary | |
Murine Ly-6C, Mr 18 kDa | |
Store at 4C in the dark. | |
IgG2c |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction