Learn More
anti-Ly-6G6C, Biotin, Clone: RB6-8C5, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128168
Catalog No. NBP128168
Add to cart
Description
Description
Ly-6G/Ly-6C Monoclonal antibody specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay.
Specifications
Specifications
Ly-6G/Ly-6C
RB6-8C5
Biotin
PBS with 0.05% Sodium Azide
LY6G
Normal mouse bone marrow.
Protein A or G purified
RUO
Primary
546644
Mouse
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro assay
0.5mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay
Purified
Rat
IgG2b Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Studies indicate that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also reacts with an additional subpopulation of Ly-6C high cells.