Learn More
anti-MAdCAM-1, Clone: MECA-367, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128146
Catalog No. NBP128146
Add to cart
Description
Description
MAdCAM-1 Monoclonal antibody specifically detects MAdCAM-1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Block/Neutralize.
Specifications
Specifications
MAdCAM-1
MECA-367
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
hMAdCAM-1, MACAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1
Rat
IgG2a Kappa
0.5 mg
Store at 4C. Do not freeze.
Monoclonal
Murine MAdCAM-1 (Mr 58-88 kDa)
Blocking Assay, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
0.5 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1 ug/ml, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing 50-200 molar excess
Purified
MADCAM1
Endothelial cells isolated from BALB/c mesenteric and peripheral lymph nodes
Protein A or G purified
RUO
Primary
8174
Mouse