Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti- MBL-2, Clone: 16AB, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP151146

Catalog No. NBP151146


Add to cart

Description

Description

MBL-2 Monoclonal antibody specifically detects MBL-2 in Mouse samples. It is validated for Western Blot, Immunoassay.
Specifications

Specifications

MBL-2
16AB
Western Blot 1:100-1:2000, Immunoassay
Purified
Mbl2
MBL is an oligomeric lectin that recognizes carbohydrates as mannose and N-acetylglucosamine on pathogens.
Protein A or G purified
RUO
Primary
17195
Immunoassay, Western Blot
Unconjugated
0.2 um filtered solution in PBS and 0.1% BSA with 0.02% Sodium Azide
L-MBP, mannose-binding protein C, mannose-binding lectin (protein C) 2, MBL, MBL-C, MBP-C
Rat
IgG2a
0.05 mg
Store at 4C. Do not freeze.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only