Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse IgA, Clone: LO-MA-7, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142335

Catalog No. NBP142335


Add to cart

Description

Description

Mouse IgA Monoclonal antibody specifically detects Mouse IgA in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

Mouse IgA
LO-MA-7
Unconjugated
PBS with 0.1% Sodium Azide
Rat
IgM Kappa
0.2 ml
Store at 4C. Do not freeze.
Monoclonal
Mouse
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
1.0 mg/mL
ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
The antibody reacts with the Mouse IgA antigen.
IgM purified
RUO
Primary
Mouse alpha heavy chain of immunoglobulin (determined by immunodot), avidity on IgA: 4.8 * 109 M-1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only