Learn More
anti-Mouse IgG1, Clone: LO-MG1-2, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142333
Catalog No. NBP142333
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IgG1 Monoclonal antibody specifically detects IgG1 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
IgG1
LO-MG1-2
Unconjugated
PBS with 0.1% Sodium Azide
Rat
IgG1 Kappa
0.2 ml
Store at 4C. Do not freeze.
Monoclonal
Mouse
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
1.0mg/mL
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
The antibody reacts with the Mouse IgG1 antigen.
Protein A or G purified
RUO
Secondary
Mouse gamma 1 heavy chain of immunoglobulin (determined by immunodot). No cross reactivity with other animals.