Learn More
anti-Mouse IgG2a, Clone: LO-MG2a-2, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142332
Catalog No. NBP142332
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IgG2a Monoclonal antibody specifically detects IgG2a in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
IgG2a
LO-MG2a-2
Unconjugated
Purified
The antibody reacts with the Mouse IgG2a antigen.
Protein A or G purified
RUO
Secondary
Mouse Gamma 2a Heavy Chain of Immunoglobulin (determined by immunodot). Avidity on IgG2a: 7 * 109 M-1. No cross reactivity with other animals.
Immunohistochemistry, Immunohistochemistry (Frozen)
1.0mg/mL
Immunohistochemistry, Immunohistochemistry-Frozen
Rat
IgG2a Kappa
0.2 ml
Store at 4C. Do not freeze.
Monoclonal
Mouse