Learn More
anti-Mouse IgG2b, Clone: LO-MG2b-2, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142331
Catalog No. NBP142331
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IgG2b Monoclonal antibody specifically detects IgG2b in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
IgG2b
LO-MG2b-2
Unconjugated
Purified
The antibody reacts with the Mouse IgG2b antigen.
Protein A or G purified
RUO
Secondary
Mouse Gamma 2b Heavy Chain of Immunoglobulin (determined by immunodot). Avidity on IgG2b: 1 * 1010 M-1. No cross reactivity with other animals.
ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
1.0mg/mL
ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Rat
IgG1 Kappa
0.2 ml
Store at 4C. Do not freeze.
Monoclonal
Mouse