Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-MRP6, Clone: M6II-7, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142342

Catalog No. NBP142342


Add to cart

Description

Description

MRP6 Monoclonal antibody specifically detects MRP6 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

MRP6
M6II-7
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Supernatant
ABCC6
The antibody reacts with the MRP6 antigen.
Tissue culture supernatant
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
M6II-7 reacts with MRP6, a 190-200 kD transmembrane protein that is related to the multidrug resistance related protein MRP. Mutations in the MRP6 gene are responsible for the connective tissue disorder Pseudoxanthoma elasticum (PXE). M6II-7 was raised against a bacterial fusion protein of human MRP6, containing amino acids 764-964, spanning the putative 12th transmembrane region as well as predicted internal and external regions of the protein. M6II-7 did not cross-react with the human MDR1, MRP1, MRP2, MRP3, MRP4, MRP5 gene products.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Unconjugated
1.0% BSA with 0.1% Sodium Azide
Anthracycline resistance-associated protein, ARAPXE, ATP-binding cassette, sub-family C (CFTR/MRP), member 6, EST349056, MLP1ABC34, MOATE, MOAT-E, MRP6ATP-binding cassette sub-family C member 6, multidrug resistance-associated protein 6, Multi-specific organic anion transporter E, pseudoxanthoma elasticum, PXE1, URG7
Rat
IgG2 Alpha
0.2 ml
Cancer
Primary
368
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only