Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-P2X7/P2RX7, PerCP, Clone: Hano43, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP140894PCP

Catalog No. NBP140894P


Add to cart

Description

Description

P2X7/P2RX7 Monoclonal antibody specifically detects P2X7/P2RX7 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

P2X7/P2RX7
Hano43
Flow Cytometry, Immunocytochemistry/Immunofluorescence
Purified
P2RX7
A P2X7-expression construct and a final boost with P2X7-transfected HEK cells.
Protein G purified
RUO
Primary
5027
Mouse
Flow Cytometry, Immunocytochemistry, Immunofluorescence
PerCP
PBS with 0.05% Sodium Azide
ATP receptor, MGC20089, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A
Rat
IgG2b
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognises the P2X purinoceptor 7, also known as P2X7. The P2X7 ATP receptor has a distinctive long C-terminal tail with multiple potential protein and lipid interaction motifs and is highly polymorphic. It is a cation selective ion channel that opens up on binding of extracellular ATP. Sustained activation by extracellular ATP results in the formation of a reversible pore in the plasma membrane that is permeable to hydrophilic solutes of up to 900 Da. Once a pore is opened massive upset of cytoplasmic ion homeostasis occurs and the pore stays open as long as it is bound by ATP. Should ATP stimulation continue the cell will become irreversibly damaged and die. P2X7 plays a key role in the maturation and release of IL-1 and other IL-1 family members during inflammation. As such, P2X7 blockers might be useful as anti-inflammatory agents.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only