Learn More
anti-TCP1-beta, Clone: PK/8/4/4i/2f, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11011450
Catalog No. NB11011450
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
TCP1-beta Monoclonal antibody specifically detects TCP1-beta in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
TCP1-beta
PK/8/4/4i/2f
Unconjugated
PBS (pH 7.2), 10mM Sodium Phosphate 50mM NaCl and 0.1mM PMSF with No Preservative
Cctb, CCT-beta, CCTBPRO1633,99D8.1, chaperonin containing t-complex polypeptide 1, beta subunit, chaperonin containing t-complex polypeptide 1, subunit 2, chaperonin containing TCP1, subunit 2 (beta), MGC142074, MGC142076, T-complex protein 1 subunit beta, T-complex protein 1, beta subunit, TCP-1-beta
Rat
IgG2b
0.05 mg
Cell Cycle and Replication, Membrane Trafficking and Chaperones
Primary
10576
Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Hamster, Monkey, Rabbit, Sheep
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
0.34 mg/mL
Western Blot 1:100-1:2000, Immunohistochemistry 10 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Affinity Purified
CCT2
TCP1 beta peptide conjugated to purified tuberculin protein derivative (PPD) (Mouse).
Protein A or G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This does not react with human Hsp60 nor with E. coli GroEL proteins.