Learn More
anti-TfR (Transferrin R), FITC, Clone: RI7217, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128017
Catalog No. NBP128017
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
TfR (Transferrin R) Monoclonal antibody specifically detects TfR (Transferrin R) in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, In vitro assay, Immunocytochemistry.
Specifications
Specifications
TfR (Transferrin R)
RI7217
FITC
PBS with 0.05% Sodium Azide
Purified
TFRC
DMSO induced Friend erythroleukemia 745.6
Protein A or G purified
RUO
Primary
7037
Mouse
Western Blot, Flow Cytometry, Immunoprecipitation, In vitro assay, Immunocytochemistry
0.5mg/mL
Western Blot 1:100-1:2000, Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500, In vitro assay, Immunocytochemistry
P02786
CD71 antigen, CD71TRFR, p90, T9, TfR, TFR1, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Rat
IgG2a Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
CD71/transferrin receptor, Mr 180-190 kDa (90-95 kDa under reducing conditions)