Learn More
anti-TLR9, Alexa Fluor 700, Clone: M9.D6, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP143141AF700
Catalog No. NBP143141Z
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
TLR9
M9.D6
Western Blot, Flow Cytometry, Flow (Intracellular)
Affinity Purified
TLR9
The immunogen for this antibody was Tolllike receptor 9.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
CD289, CD289 antigen, toll-like receptor 9
Rat
IgG2a
0.1 ml
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary
54106