Learn More
anti-TLR9, DyLight 405, Clone: eB72-1665, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143140V
Catalog No. NBP143140V
Add to cart
Description
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
TLR9
eB72-1665
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
Q9NR96
CD289, CD289 antigen, toll-like receptor 9
Rat
IgG2a Kappa
0.1 ml
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary
54106
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
TLR9
The immunogen for this antibody was Tolllike receptor 9.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Human