Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-TLR9, PE, Clone: M9.D6, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP143141PE

Catalog No. NBP143141PE

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TLR9
M9.D6
Flow Cytometry, Flow (Intracellular)
Affinity Purified
TLR9
The immunogen for this antibody was Tolllike receptor 9.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
CD289, CD289 antigen, toll-like receptor 9
Rat
IgG2a
0.1 ml
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary
54106
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only