Learn More
TREM1 Rat anti-Mouse, DyLight 405, Clone: L5-B8.2A12.3A12, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP140907V
Catalog No. NBP140907V
Add to cart
Description
Description
TREM1 Monoclonal specifically detects TREM1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
Specifications
TREM1
L5-B8.2A12.3A12
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Purified
TREM1
Mouse TREM-1 Fc fusion protein.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
CD354 antigen, TREM-1Triggering receptor expressed on monocytes 1, triggering receptor expressed on myeloid cells 1, triggering-receptor TREM1
Rat
IgG2a
0.25 ml
Signal Transduction
Primary
54210