Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat TrkA Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 18 publications
Supplier: R&D Systems AF1056
This item is not returnable.
View return policy
Description
TrkA Polyclonal specifically detects TrkA in Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TrkA | |
Polyclonal | |
Unconjugated | |
P35739 | |
NTRK1 | |
Mouse myeloma cell line NS0-derived recombinant rat TrkA Ala33-Pro418 Accession # P35739 | |
100 μg | |
Primary | |
Detects rat TrkA in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human TrkA and less than 1% cross-reactivity with recombinant mouse (rm) TrkB and rmTrkC is observed. | |
Rat | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
DKFZp781I14186, EC 2.7.10, EC 2.7.10.1, MTChigh affinity nerve growth factor receptor, Neurotrophic tyrosine kinase receptor type 1, neurotrophic tyrosine kinase, receptor, type 1, NTRK1, NTRK-1, p140-TrkA, TRK1-transforming tyrosine kinase protein, Trk-A, TRKAOncogene TRK, TRKTRK1, tyrosine kinase receptor A | |
Goat | |
Affinity Purified | |
RUO | |
4914 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction