Learn More
VEGFR3/Flt-4 Rat anti-Mouse, Reptile, DyLight 755, Clone: AFL4, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143259IR
Catalog No. NBP143259IR
Add to cart
Description
Description
VEGFR3/Flt-4 Monoclonal antibody specifically detects VEGFR3/Flt-4 in Mouse, Reptile samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications
Specifications
VEGFR3/Flt-4
AFL4
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Affinity Purified
FLT4
The immunogen for this antibody was VEGFR3.
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4
Rat
IgG2a Kappa
0.1 ml
Angiogenesis, Cancer, Hypoxia, Protein Kinase
Primary
2324