Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAX Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124451

Catalog No. PIPA124451


Add to cart

Description

Description

PA1-24451 detects RAX in Human samples. PA1-24451 has been successfully used in Western Blot procedures. PA1-24451 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human RAX. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Retinal Homeobox Protein Rx (RAX) is a member of homeobox family of transcription factors. Mutations mouse and fish RAX lead to defects in retinal development and result in animal models of anophthalmia.
Specifications

Specifications

RAX
Polyclonal
Unconjugated
RAX
Liquid
RAX
Synthetic peptide region of human RAX.
IgG fraction
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q9Y2V3
MCOP3, RX
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
30062
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit