Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-RB IGG (H+L) AB DL488  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300339
Affinity purified polyclonal antibody to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 488. Product is in lyophilized form.
Catalog No. 50-669-49