Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-RB IGG (H+L) AB HSA DL488  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52300386
Affinity purified polyclonal antibody to Rabbit IgG, both heavy and light chains (whole IgG) made in Goat and labeled with DyLight 488. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form. The conjugate is designed for use in a variety of immunofluorescense assays, including immunohistology, flow cytometry, Western blotting, microarrays and Fluorophore-Linked Immunosorbent Assays (FLISA).
Catalog No. 50-670-80