Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-RB IGG (H+L) AB RESERVEAP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200465
Affinity purified polyclonal antibody to Rabbit IgG heavy and light chains (whole IgG) made in Goat and labeled with alkaline phosphatase. ReserveAP conjugates represent the latest advances in KPL's conjugation chemistry and are more sensitive than KPL's standard AP conjugates. Product is in liquid form. Each lot is tested to assure specificity and lot-to-lot consistency using KPL's in-house ELISA assay.
Catalog No. 50-673-96