anti-rba-1, Polyclonal, Novus Biologicals™

Manufacturer: Novus BiologicalsSupplier Diversity Partner 4875000201ML

 View more versions of this product

Catalog No. 48-750-00201M

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Disclaimers:

For Research Use Only


    Specifications

    Format Affinity Purified
    Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
    Regulatory Status RUO
    Monoclonal or Polyclonal Polyclonal
    Gene ID (Entrez) 172801
    Concentration 1mg/mL
    Cross Reactivity C. elegans
    Antigen rba-1
    Host Species Rabbit
    Applications ELISA,Immunocytochemistry,Immunofluorescence
    Immunogen In vivo generated recominant protein fragment
    Quantity 0.1mL
    Conjugate Unlabeled
    Primary or Secondary Primary
    Test Specificity Caenorhabditis elegans.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit