anti-rba-1, Polyclonal, Novus Biologicals™

Manufacturer: Novus BiologicalsSupplier Diversity Partner 48750002

 View all options for this product

Catalog No. 48-750-002

  Check Availability
Add to Cart


For Research Use Only


    Antigen rba-1
    Applications ELISA,Immunocytochemistry,Immunofluorescence
    Concentration 1mg/mL
    Conjugate Unlabeled
    Cross Reactivity C. elegans
    Format Affinity Purified
    Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
    Host Species Rabbit
    Immunogen In vivo generated recominant protein fragment
    Quantity 0.05mL
    Regulatory Status RUO
    Primary or Secondary Primary
    Monoclonal or Polyclonal Polyclonal
    Gene ID (Entrez) 172801
    Test Specificity Caenorhabditis elegans.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit