Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527711

Catalog No. PIPA527711


Add to cart

Description

Description

PA5-27711 targets RBBP5 in IF and WB applications and shows reactivity with Human samples. The PA5-27711 immunogen is recombit fragment corresponding to a region within amino acids 20 and 223 of Human RBBP5. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that bind directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B.
Specifications

Specifications

RBBP5
Polyclonal
Unconjugated
RBBP5
Liquid
RBBP5
Recombit fragment corresponding to a region within amino acids 20 and 223 of Human RBBP5
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.11 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
Q15291
RBQ3, SWD1, RBBP-5, SWD1, Set1c WD40 repeat protein, homolog, retinoblastoma-binding protein 5, retinoblastoma-binding protein RBQ-3
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5929
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit