Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CtIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520963
Description
A suggested positive control is mouse spleen tissue lysate. PA5-20963 can be used with blocking peptide PEP-1077.
CtIP Polyclonal antibody specifically detects CtIP in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
CtIP | |
Polyclonal | |
Unconjugated | |
Rbbp8 | |
9930104E21Rik; COM1; CtBP-interacting protein; CtIP; DNA endonuclease RBBP8; JWDS; RB binding protein 8, endonuclease; RBBP8; RBBP-8; retinoblastoma binding protein 8; retinoblastoma-binding protein 8; Retinoblastoma-interacting protein and myosin-like; RGD1308872; RIM; SAE2; SCKL2; Sporulation in the absence of SPO11 protein 2 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
225182, 291787, 5932 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
B1WC58, Q80YR6, Q99708 | |
Rbbp8 | |
A 14 amino acid peptide near the center of human RBBP8. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction