Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

retinoblastoma binding protein 9, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human RBBP9 protein.

Manufacturer:  Abnova Corporation H00010741B01

Catalog No. 89-007-046


Add to cart

Description

Description

The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate. [provided by RefSeq

Sequence: MASPSKAVIVPGNGGGDVTTHGWYGWVKKELEKIPGFQCLAKNMPDPITARESIWLPFMETELHCDEKTIIIGHSSGAIAAMRYAETHRVYAIVLVSAYTSDLGDENERASGYFTRPWQWEKIKANCPYIVQFGSTDDPFLPWKEQQEVADRLETKLHKFTDCGHFQNTEFHELITVVKSLLKVPA
Specifications

Specifications

retinoblastoma binding protein 9
Unconjugated
No additive
NM_006606.2
BOG|MGC9236|RBBP10
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
10741
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human RBBP9 protein.
RBBP9
Serum
RBBP9
RBBP9 (NP_006597.2, 1 a.a. ~ 186 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit