Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RBM3, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136980

Catalog No. NBP136980


Add to cart

Description

Description

RBM3 Polyclonal antibody specifically detects RBM3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications

Specifications

RBM3
0.5 mg/mL
Western Blot 0.01 - 0.03 ug/ml, Peptide ELISA Detection limit 1:2000
NP_006734.1
IS1-RNPL, putative RNA-binding protein 3, RNA binding motif (RNP1, RRM) protein 3, RNA binding motif protein 3, RNA-binding motif protein 3, RNPL
Goat
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific to the C Terminus. Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Pig and Cow.
ELISA, Western Blot
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
RBM3
Peptide with sequence C-DRYSGGNYRDNYDN corresponding to C-Terminus according to NP_006734.1.
Immunogen affinity purified
RUO
Primary
5935
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only