Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514290

Catalog No. PIPA514290


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

RBM3 is a member of the glycine-rich RNA-binding protein family and a protein with one RNA recognition motif (RRM) domain. Expression of this protein is induced by cold shock and low oxygen tension.
Specifications

Specifications

RBM3
Polyclonal
PBS with 0.09% sodium azide
P98179, O89086
Putative RNA-binding protein 3; RNA-binding motif protein 3; RNPL; RNPL; RBM3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
19652, 5935
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RBM3
Liquid
RBM3
KLH conjugated synthetic peptide between 55-84 amino acids from the central region of human RBM3
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit