Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521755

Catalog No. PIPA521755


Add to cart

Description

Description

PA5-21755 targets RBM4 in ICC, IF and WB applications and shows reactivity with Human samples. The PA5-21755 immunogen is synthetic peptide contain a sequence corresponding to a region within amino acids 304 and 316 (Q9BWF3) of RBM4. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

RBM4
Polyclonal
Unconjugated
RBM4
Liquid
RBM4
Synthetic peptide contain a sequence corresponding to a region within amino acids 304 and 316 (Q9BWF3) of RBM4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
Q9BWF3
LARK, RBM4A, ZCCHC21, ZCRB3A, RNA-binding motif protein 4a, RNA-binding protein 4, lark homolog, transcriptional coactivator CoAZ, zinc finger CCHC-type and RNA binding motif 3A
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5936
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit