Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM43 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524106

Catalog No. PIPA524106

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

RBM43
Polyclonal
PBS with 0.09% sodium azide
Q6ZSC3
C2orf38, RNA-binding motif protein 43, RNA-binding protein 43
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
375287
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
RBM43
Liquid
RBM43
KLH conjugated synthetic peptide between 21-49 amino acids from the N-terminal region of human RBM43
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit