Learn More
RBMS1 Mouse anti-Human, Clone: 3F2-2G9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA519797
Catalog No. PIMA519797
Add to cart
Description
Description
Peptide Sequence: MGKVWKQQMY PQYATYYYPQ YLQAKQSLVP AHPMAPPSPS TTSSNNNSSS SSNSGWDQLS KTNLYIRGLP PHTTDQDLVK LCQPYGKIVS TKAILDKTTN KCKGYGFVDF DSPAAAQKAV SALKASGVQA QMAKQQEQDP TNLYISNLPL SMDEQELENM LKPFGQVIST RILRDSSGTS RGVGFARMES TEKCEAVIGH FNGKFIKTPP GVSAPTEPLL CKFADGGQKK RQNPNKYIPN GRPWHREGEA GMTLTYDPTT AAIQNGFYPS PYSIATNRMI TQTSITPYIA SPVSAYQVQS PSWMQPQPYI LQHPGAVLTP SMEHTMSLQP ASMISPLAQQ MSHLSLGSTG TYMPATSAMQ GAYLPQYAHM QTTAVPVEEA SGQQQVAVET SNDHSPYTFQ PNK
This gene encodes a member of a small family of proteins which bind single stranded DNA/RNA. These proteins are characterized by the presence of two sets of ribonucleoprotein consensus sequence (RNP-CS) that contain conserved motifs, RNP1 and RNP2, originally described in RNA binding proteins, and required for DNA binding. These proteins have been implicated in such diverse functions as DNA replication, gene transcription, cell cycle progression and apoptosis. Several transcript variants, resulting from alternative splicing and encoding different isoforms, have been described. A pseudogene for this locus is found on chromosome 12.
Specifications
Specifications
RBMS1
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P29558
C2orf12, HCC-4, MSSP, MSSP-1, MSSP-2, MSSP-3, SCR2, YC1, RNA-binding motif, single-stranded-interacting protein 1, c-myc gene single strand binding protein 2, cervical cancer oncogene 4, single-stranded DNA-binding protein MSSP-1, suppressor of CDC2 with RNA-binding motif 2, suppressor of cdc 2 (cdc13) with RNA bind
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
5937
ELISA, Western Blot
3F2-2G9
Unconjugated
RBMS1
Liquid
RBMS1
RBMS1 (AAH12993, 1 a.a. ∽ 403 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human