Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBP Mouse, Unlabeled, Clone: 42, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610237

Catalog No. BDB610237


Add to cart

Description

Description

Retinol, a metabolic precursor of retinal and retinoic acid, is transported in plasma bound to a carrier protein known as retinol-binding protein (RBP). RBP is the only retinoid-specific binding protein that has been identified in plasma. The presence of RBP increases the stability of retinol as it is transported from its stores to target cells. In plasma, the 21 kDa RBP forms a tight complex with thyroxine-binding transthyretin (TTR), a homotetramer of about 55 kDa. Association of RBP with TTR increases the stability of the retinol-RBP complex. It is believed that this association prevents filtration of the relatively small RBP through the kidney glomeruli. Once retinol is delivered to its target cells, the resulting retinol-free RBP has a reduced affinity for TTR and thus can be selectively cleared from the circulation by filtration through kidney glomeruli. The three dimensional structures of human and bovine RBP have been compared and appear to be very similar.

Specifications

Specifications

RBP
42
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit