Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBPJ Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA522226
Description
Recommended positive controls: 293T, A431, HeLa, Molt-4, Raji. Predicted reactivity: Mouse (97%), Rat (98%), Xenopus laevis (93%), Rhesus Monkey (99%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RBPJ Polyclonal antibody specifically detects RBPJ in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
RBPJ | |
Polyclonal | |
Unconjugated | |
RBPJ | |
AI843960; AOS3; CBF1; CBF-1; csl; H-2K binding factor-2; Igkjrb; Igkjrb1; Igkrsbp; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; KBF2; Rbpj; RBP-J; RBP-J kappa; RBPJK; RBP-JK; rbp-jkappa; RBPSUH; recombination signal binding protein for immunoglobulin kappa J region; recombining binding protein suppressor of hairless; recombining binding protein suppressor of hairless-like; recombining binding protein suppressor of hairless-like protein; renal carcinoma antigen NY-REN-30; SUH; suppressor of hairless homolog; Unknown (protein for MGC:128782) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
19664, 3516 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.75 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P31266, Q06330 | |
RBPJ | |
Recombinant fragment corresponding to a region within amino acids 258 and 500 of RBP-Jkappa (Uniprot ID#Q06330) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction