Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526563

Catalog No. PIPA526563

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

RBP2 is an abundant protein present in the small intestinal epithelium. It is thought to participate in the uptake and/or intracellular metabolism of vitamin A. Vitamin A is a fat-soluble vitamin necessary for growth, reproduction, differentiation of epithelial tissues, and vision. RBP2 may also modulate the supply of retinoic acid to the nuclei of endometrial cells during the menstrual cycle.
Specifications

Specifications

RBP2
Polyclonal
PBS with 0.09% sodium azide
P50120
CRABP-II, CRBP2, CRBPII, RBPC2, CRBP-II, cellular retinol-binding protein II, retinol-binding protein 2, retinol-binding protein 2, cellular
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5948
Western Blot
Unconjugated
RBP2
Liquid
RBP2
KLH conjugated synthetic peptide between 56-84 amino acids from the central region of human RBP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit