Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RCBTB2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133512

Catalog No. NBP133512


Add to cart

Description

Description

RCBTB2 Polyclonal antibody specifically detects RCBTB2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RCBTB2
0.32mg/mL
Western Blot 1:500-1:3000
Affinity Purified
RCBTB2
Recombinant protein encompassing a sequence within the center region of human RCBTB2. The exact sequence is proprietary.
60 kDa
0.1 ml
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
CHC1-L, CHC1LRCC1-like G exchanging factor RLG, Chromosome condensation 1-like, RCC1 and BTB domain-containing protein 2, RCC1-like G exchanging factor, regulator of chromosome condensation (RCC1) and BTB (POZ) domain containingprotein 2, regulator of chromosome condensation and BTB domain containing protein 2, Regulator of chromosome condensation and BTB domain-containing protein 2, RLG
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
1102
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only