Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCN3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525688

Catalog No. PIPA525688


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Calcipressin 3 inhibits calcineurin dependent transcriptional responses by binding to the catalytic domain of calcineurin A, and may play a role during central nervous system development. Highest expression occurs in heart, skeletal muscle kidney, liver and peripheral blood leukocytes.
Specifications

Specifications

RCN3
Polyclonal
PBS with 0.09% sodium azide
Q96D15, Q8BH97
RLP49, EF-hand calcium-binding protein RLP49, reticulocabin, reticulocalbin-3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
52377, 57333
Flow Cytometry, Western Blot
Unconjugated
RCN3
Liquid
RCN3
KLH conjugated synthetic peptide between 134-163 amino acids from the central region of human RCN3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit