Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RDH16 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525370

Catalog No. PIPA525370

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

Retinol dehydrogenase 16 (RDH16) is an oxidoreductase with a preference for NAD. It oxidizes all-trans-retinol and 13-cis-retinol to the corresponding aldehydes. RDH16 has higher activity towards CRBP-bound retinol than with free retinol. It oxidizes 3-alpha-hydroxysteroids and also oxidizes androstanediol and androsterone to dihydrotestosterone and androstanedione. RDH16 can also catalyze the reverse reaction.
Specifications

Specifications

RDH16
Polyclonal
PBS with 0.09% sodium azide
O75452
RODH-4, SDR9C8, microsomal NAD+-dependent retinol dehydrogenase 4, retinol dehydrogenase 16, retinol dehydrogenase 16 (all-trans and 13-cis), short chain dehydrogenase/reductase family 9C, member 8, sterol/retinol dehydrogenase
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
8608
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RDH16
Liquid
RDH16
KLH conjugated synthetic peptide between 3-31 amino acids from the N-terminal region of human RDH16
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit