Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Recoverin Mouse, Unlabeled, Clone: S36-208, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564905

Catalog No. BDB564905

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Recoverin
S36-208
Unconjugated
Purified
Human Recoverin Recombinant Protein
0.1mg
Store undiluted at 4°C
Monoclonal
Western Blot
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1
RUO
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.