Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RECQL5, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP140340

Catalog No. NBP140340


Add to cart

Description

Description

RECQL5 Polyclonal antibody specifically detects RECQL5 in Human samples. It is validated for Western Blot, Immunoprecipitation, Tissue Culture Substratum.
Specifications

Specifications

RECQL5
1.0 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 5-15 ug/mg lysate, Tissue Culture Substratum
O94762
DNA helicase, RecQ-like type 5, EC 3.6.1, EC 3.6.4.12, FLJ90603, RecQ protein-like 5ATP-dependent DNA helicase Q5, RecQ5, RECQ5RecQ protein 5
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation, Tissue Culture Substratum, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
RECQL5
The immunogen for this product maps to a region between residue 450 and 500 of human RecQ protein-like 5 using the numbering given in entry NP_004250.4 (GeneID 9400).
Immunogen affinity purified
RUO
Primary
9400
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only