Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REEP4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524474

Catalog No. PIPA524474

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May enhance the cell surface expression of odorant receptors.
Specifications

Specifications

REEP4
Polyclonal
PBS with 0.09% sodium azide
Q9H6H4, Q8K072
C8orf20, receptor expression enhancing protein 4, receptor expression-enhancing protein 4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
72549, 80346
Western Blot
Unconjugated
REEP4
Liquid
REEP4
KLH conjugated synthetic peptide between 228-257 amino acids from the C-terminal region of human REEP4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit