Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REG3A, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526219

Catalog No. PIPA526219

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

REG3A encodes a pancreatic secretory protein that may be involved in cell proliferation or differentiation. It has similarity to the C-type lectin superfamily. The enhanced expression of this protein is observed during pancreatic inflammation and liver carcinogenesis.

Specifications

Specifications

REG3A
Polyclonal
PBS with 0.09% sodium azide
Q06141
HIP, PAP, PAP-H, PAP1, PBCGF, REG-III, REG3, PAP homologous protein, REG-3-alpha, hepatocarcinoma-intestine-pancreas, human proislet peptide, pancreatic beta cell growth factor, pancreatitis-associated protein 1, proliferation-inducing protein 34, proliferation-inducing protein 42, reg III-alpha, regeneratin
Rabbit
IgG
400μL
Store at 2°-8°C for up to 6 months. For long term storage store at -20°C
Primary
5068
Flow Cytometry, Western Blot
Unconjugated
REG3A
Purified
REG3A
KLH conjugated synthetic peptide between 12-39 amino acids from the N-terminal region of human REG3A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit