Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reg3D Rat anti-Mouse, Clone: 518818, Invitrogen™

Rat Monoclonal Antibody

Manufacturer:  Invitrogen MA524226

Catalog No. PIMA524226


Add to cart

Description

Description

In direct ELISAs, no cross-reactivity with recombit human (rh) Reg1A, 1B, recombit mouse (rm) Reg1, 2, 3A, 3B, 3G, 4, or recombit rat Reg3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

Reg3D
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Lyophilized
Reg3d
E. coli-derived recombit mouse Reg3D Glu27-Gly175
Protein A/G
RUO
Antibody
Monoclonal
Mouse
Immunohistochemistry (Frozen)
518818
Unconjugated
Reg3d
INGAPrP
Rat
IgG2A
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
30053
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit