Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REG1A Rabbit anti-Human, Non-human primate, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523314

Catalog No. PIPA523314


Add to cart

Description

Description

Regenerating Islet-derived 1, also known as REG1, is a novel protein encoded by a multigene family comprising of REG and REG-related genes known to be involved in regeneration. REG1A and REG1B are the proteins encoded by these REG families of genes and belong to the pancreatic stone protein family. They are comprised of 166 amino acids and differed by only 22 amino acids with a C-type lectin domain. Known to be identical to that of pancreatic stone protein, REG proteins may be involved in the expansion of beta-cell mass during regeneration as well as in the maintece of normal beta-cell function. These may act as inhibitors of spontaneous calcium carbonate precipitation and may also be associated with neuronal sprouting in brain, and with brain and pancreas (islet) regeneration. Expression of REG proteins is common in pancreas and liver. 
Specifications

Specifications

REG1A
Polyclonal
Unconjugated
REG1A
Liquid
REG1A
Synthetic peptide (21 QGQESQTELPNPRISC 36) of Regenerating Islet-derived 1 protein.
Protein G
RUO
Antibody
Polyclonal
Human, Non-human Primate
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P05451
ICRF, P19, PSP, PSPS, PSPS1, PTP, REG, REG-1-alpha, islet cells regeneration factor, islet of langerhans regenerating protein, lithostathine 1 alpha, lithostathine-1-alpha, pancreatic stone protein, secretory, pancreatic thread protein, protein-X, regenerating islet-derived protein 1-alpha, regenerating protein I alpha
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
5967
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit