Learn More
anti-RelA/NFkB p65 (p Ser276), Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP177827
Catalog No. NBP177827
Add to cart
Description
Description
RelA/NFkB p65 p Ser276 Polyclonal antibody specifically detects RelA/NFkB p65 p Ser276 in Human samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
RelA/NFkB p65 (p Ser276)
1 mg/ml
Western Blot 1:1000, ELISA 1:5000-1:25000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500
Q99496
MGC131774, nf kb p65, NF kB p65 phospho, NFkB p65, NF-kB p65, NFkB p65 phospho, NF-kB p65 phospho, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, phospho NF kB p65, phospho NFkB p65, phospho NF-kB p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Rabbit
IgG
0.1 mg
Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary
5970
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Delipidation and Defibrination
RELA
RelA/NFkB p65 peptide corresponding to a region near [p Ser276] of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH).
Delipidation and Defibrination
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This phospho specific polyclonal antibody is specific for phosphorylated pS276 human p65. Reactivity with non-phosphorylated p65 is minimal.