Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-RelA/NFkB p65 (p Ser529), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP177808

Catalog No. NBP177808


Add to cart

Description

Description

RelA/NFkB p65 p Ser529 Polyclonal antibody specifically detects RelA/NFkB p65 p Ser529 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

RelA/NFkB p65 (p Ser529)
75 mg/mL
Western Blot 1:500-1:3000, ELISA 1:5000-1:25000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:10-1:500
Q04206
MGC131774, nf kb p65, NF kB p65 phospho, NFkB p65, NF-kB p65, NFkB p65 phospho, NF-kB p65 phospho, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, phospho NF kB p65, phospho NFkB p65, phospho NF-kB p65, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Rabbit
Serum
0.1 ml
Apoptosis, Cancer, Cell Biology, Immune System Diseases, Immunology, Innate Immunity, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction, Transcription Factors and Regulators
Primary
5970
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Antiserum with 0.1% Sodium Azide
Antisera
RELA
RelA/NFkB p65 peptide corresponding to a region near [p Ser529] of the human protein conjugated to Keyhole Limpet Hemocyanin (KLH).
Delipidation and Defibrination
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
This phospho specific polyclonal antibody is specific for phosphorylated pS529 human p65. Reactivity with non-phosphorylated p65 is minimal.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only