Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-RENI SALMONINARUM AB  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 53100336
Affinity purified polyclonal antibody to Renibacterium salmoninarum made in Goat. Isolated from a serum pool of goats immunized with heat killed whole cells of Renibacterium salmonarium. The antibody is highly specific for Renibacterium salmoninarum.
Catalog No. 50-668-67