Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ATP6IP2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA528440
Description
PA5-28440 targets Renin Receptor in WB applications and shows reactivity with Human samples. The PA5-28440 immunogen is synthetic peptide corresponding to a region within amino acids 287 and 350 of Human Renin Receptor.
This gene encodes a protein that is associated with adenosine triphosphatases (ATPases). Proton-translocating ATPases have fundamental roles in energy conservation, secondary active transport, acidification of intracellular compartments, and cellular pH homeostasis. There are three classes of ATPases- F, P, and V. The vacuolar (V-type) ATPases have a transmembrane proton-conducting sector and an extramembrane catalytic sector. The encoded protein has been found associated with the transmembrane sector of the V-type ATPases.
Specifications
ATP6IP2 | |
Polyclonal | |
Unconjugated | |
O75787 | |
ATP6AP2 | |
Synthetic peptide corresponding to a region within amino acids 287 and 350 of Human Renin Receptor | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
ATP6AP2 | |
APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, M8-9, MRXE, MSTP009, XMRE, ATPase H(+)-transporting lysosomal-interacting protein 2, ATPase, H+ transporting, lysosomal (vacuolar proton pump) membrane sector associated protein M8-9, ATPase, H+ transporting, lysosomal interacting protein 2, ER-localized type I transmembrane adaptor, N | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10159 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction